Learn More
Sigma Aldrich Fine Chemicals Biosciences Diamond Shaped Weigh Boats | 5ml | 500ea
Catalog No. NC1750569
Encompass
These flexible polystyrene boats feature smooth surfaces to provide accurate pour-outs with minimal sample loss. Biologically inert and resistant to dilute and weak acids, aqueous solutions, alcohols, and bases.
Catalog No. NC1750569 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. HS1425AA
These flexible polystyrene boats feature smooth surfaces to provide accurate pour-outs with minimal sample loss. Biologically inert and resistant to dilute and weak acids, aqueous solutions, alcohols, and bases.