Learn More
Electron Microscopy Sciences Diamond Stylus for Microscope Slide Labelers
SDP
Catalog No. 501931662
Encompass_Preferred
This diamond stylus is perfectly suited for the Thermo Scientific PrintMate and MicroWriter, RA Lamb and TBS Shurmark microscope slide labelers. Manufactured with a consistent even coating of diamond dust. The 2.35 mm shaft is made from stainless steel.
Catalog No. 50-193-1662 Supplier Electron Microscopy Sciences Supplier No. 62538
This diamond stylus is perfectly suited for the Thermo Scientific PrintMate and MicroWriter, RA Lamb and TBS Shurmark microscope slide labelers. Manufactured with a consistent even coating of diamond dust. The 2.35 mm shaft is made from stainless steel.