Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Diamond Slurry, 1μm, 10mL

Catalog No. 50792655 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This diamond slurry is specifically formulated for polishing working electrodes.

  • Material used to polish the working electrode.
  • Particle size: 1μm.
  • Volume: 10mL.
  • Units per package: 1 pk.

Catalog No. 50-792-655 Supplier Waters Corp Supplier No. 700001955
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This diamond slurry is specifically formulated for polishing working electrodes.

  • Material used to polish the working electrode.
  • Particle size: 1μm.
  • Volume: 10mL.
  • Units per package: 1 pk.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.