Learn More
Electron Microscopy Sciences Glennel Polycrystalline Diamond Spray 0.5 µm particles 5 OZ
SDP
Catalog No. 50997357
Encompass_Preferred
Glennel Diamond Aerosol Spray provides unsurpassed cutting action and finish. The inverted nozzle provides safe and easy access for difficult to reach areas while ensuring that every diamond particle is used. Special formulations allow the carrier to evaporate quickly, leaving a very uniform dispersion of diamond. This is especially useful when an operation requires a dry polishing cloth.
Particles: 0.5 µm
Catalog No. 50-997-357 Supplier Electron Microscopy Sciences Supplier No. 5037410
Glennel Diamond Aerosol Spray provides unsurpassed cutting action and finish. The inverted nozzle provides safe and easy access for difficult to reach areas while ensuring that every diamond particle is used. Special formulations allow the carrier to evaporate quickly, leaving a very uniform dispersion of diamond. This is especially useful when an operation requires a dry polishing cloth.
Particles: 0.5 µm