Learn More
Electron Microscopy Sciences Diamond Stone, Coarse
SDP
Catalog No. 50996529
Encompass_Preferred
This patented process of heat treatment which bonds the highest quality industrial grade diamond particles in a stainless alloy to a precision matched metal substrate, has been imitated, but has never come close to being duplicated. Our stone which is set on a walnut pedestal with a cover. Coarse grit (180), size: 2"x6" (51mmx152mm)
Catalog No. 50-996-529 Supplier Electron Microscopy Sciences Supplier No. 6208252
This patented process of heat treatment which bonds the highest quality industrial grade diamond particles in a stainless alloy to a precision matched metal substrate, has been imitated, but has never come close to being duplicated. Our stone which is set on a walnut pedestal with a cover. Coarse grit (180), size: 2"x6" (51mmx152mm)