Learn More
Electron Microscopy Sciences Diamond Stone All Purpose Fine Grit
SDP
Catalog No. 50996764
Encompass_Preferred
This diamond stone has millions of cutting edges to sharpen most any tool. Many times faster than dry or oil stones. May be used as a file or a stone. It cuts in all directions. Available in all purpose fine grit.
Catalog No. 50-996-764 Supplier Electron Microscopy Sciences Supplier No. 62120
This diamond stone has millions of cutting edges to sharpen most any tool. Many times faster than dry or oil stones. May be used as a file or a stone. It cuts in all directions. Available in all purpose fine grit.