Learn More
Allied High Tech Products Diamond Suspension, Water Based Polycrystalline, 3 Micron, 16 oz. | 480mL
SDP
Catalog No. NC1809139
Encompass
Water soluble and environmentally friendly, this 99% water based formula is recommended for manual application or automatic –"spray"/"drip" dispensing systems, where lubricant is also applied. Allied diamond suspensions are specifically formulated to assure that the diamond particles remain suspended and separated throughout the stable liquid carrier. They are excellent for distributing diamond easily and uniformly over the cloth or platen surface. All 8 and 16 oz. bottles include a flip-top and trigger sprayer for dispensing. Gallons include a pre-labeled 16 oz. empty bottle for dispensing convenience.
Catalog No. NC1809139 Supplier Allied High Tech Products Supplier No. 9032020
Water soluble and environmentally friendly, this 99% water based formula is recommended for manual application or automatic –"spray"/"drip" dispensing systems, where lubricant is also applied. Allied diamond suspensions are specifically formulated to assure that the diamond particles remain suspended and separated throughout the stable liquid carrier. They are excellent for distributing diamond easily and uniformly over the cloth or platen surface. All 8 and 16 oz. bottles include a flip-top and trigger sprayer for dispensing. Gallons include a pre-labeled 16 oz. empty bottle for dispensing convenience.