Learn More
Electron Microscopy Sciences Glennel Diamond Suspension Oil Based, 3.0 µm particles 400 GR
SDP
Catalog No. 50997347
Encompass_Preferred
Glennel diamond suspension assures a consistent, uniform viscosity and dispersion of diamond without any shaking or stirring. The proprietary formulation guarantees that the particles will stay suspended indefinitely. It is ideal for automatic polishers and applications that require changing operators. Consistent, uniform viscosity and dispersion of diamond without shaking or stirring. Formulation guarantees particles will stay suspended indefinitely. Ideal for automatic polishers.
Size: 3.0 µm
Catalog No. 50-997-347 Supplier Electron Microscopy Sciences Supplier No. 5037260
Glennel diamond suspension assures a consistent, uniform viscosity and dispersion of diamond without any shaking or stirring. The proprietary formulation guarantees that the particles will stay suspended indefinitely. It is ideal for automatic polishers and applications that require changing operators. Consistent, uniform viscosity and dispersion of diamond without shaking or stirring. Formulation guarantees particles will stay suspended indefinitely. Ideal for automatic polishers.
Size: 3.0 µm