Learn More
Electron Microscopy Sciences Glennel Diamond Suspension W/S, 6.0 µm particles 400 GR
SDP
Catalog No. 50997350
Encompass_Preferred
Glennel diamond suspension assures a consistent, uniform viscosity and dispersion of diamond without any shaking or stirring. The proprietary formulation guarantees that the particles will stay suspended indefinitely. It is ideal for automatic polishers and applications that require changing operators. A versatile, water based suspension designed for most production applications. Provides excellent lubrication, uniform fast lapping action, and maximum cutting action. Compatible with all common extenders.
Size: 6.0 µm
Catalog No. 50-997-350 Supplier Electron Microscopy Sciences Supplier No. 5037271
Glennel diamond suspension assures a consistent, uniform viscosity and dispersion of diamond without any shaking or stirring. The proprietary formulation guarantees that the particles will stay suspended indefinitely. It is ideal for automatic polishers and applications that require changing operators. A versatile, water based suspension designed for most production applications. Provides excellent lubrication, uniform fast lapping action, and maximum cutting action. Compatible with all common extenders.
Size: 6.0 µm