Learn More
Electron Microscopy Sciences Diamond Tip Scriber with Retractable Tip, OAL, 60° Tip
SDP
Catalog No. 5025451
Encompass_Preferred
Sturdy scribers are in wide use throughout the laboratory. It leaves a permanent marking on virtually any surface, including metal, glass, or plastic. Each Scriber features a hexagonal aluminum pencil shaped body, mill finish, natural diamond tip and protective cap. Chrome pen with diamond mounted in a stainless steel tip. Refills. Tip finished at 60º.
Tip: 60°
Catalog No. 50-254-51 Supplier Electron Microscopy Sciences Supplier No. 70036
Sturdy scribers are in wide use throughout the laboratory. It leaves a permanent marking on virtually any surface, including metal, glass, or plastic. Each Scriber features a hexagonal aluminum pencil shaped body, mill finish, natural diamond tip and protective cap. Chrome pen with diamond mounted in a stainless steel tip. Refills. Tip finished at 60º.
Tip: 60°