Learn More
Electron Microscopy Sciences DiATOME Resharpen Trim Tool 90° Knife angles
SDP
Catalog No. 501927541
Encompass
A resharpened DiATOME knife has the same length, an identical cutting edge and carries the same guarantee of quality as a new knife. During resharpening a knife may be reworked into another type.
Catalog No. 50-192-7541 Supplier Electron Microscopy Sciences Supplier No. TTR90
A resharpened DiATOME knife has the same length, an identical cutting edge and carries the same guarantee of quality as a new knife. During resharpening a knife may be reworked into another type.