Learn More
MSC DIAMOND-TUFF MAX MAT
Catalog No. 5000793504
Encompass
This anti-fatigue mat is designed for medium-duty use in dry environments. It features a diamond plate surface, a 1 inch thickness, and beveled edges.
  • Durable diamond pattern vinyl laminate top surface for added traction
  • Dense closed cell PVC foam base provides added cushioning comfort
  • Beveled edges on all 4 sides
Catalog No. 50-007-93504 Supplier MSC Supplier No. 63962435
This anti-fatigue mat is designed for medium-duty use in dry environments. It features a diamond plate surface, a 1 inch thickness, and beveled edges.
  • Durable diamond pattern vinyl laminate top surface for added traction
  • Dense closed cell PVC foam base provides added cushioning comfort
  • Beveled edges on all 4 sides