Learn More
Grainger Micro 100 Diamond Turning Insert, CPG422, Rhombic, Uncoated, 1 ea
Catalog No. 502778668
Encompass_Preferred
This diamond turning insert is designed for precision machining operations, including turning, boring, and profiling. Featuring a rhombic diamond shape, this uncoated insert is suitable for a wide range of workpiece materials, including steel, stainless steel, cast iron, nonferrous materials, and super alloys.
  • 4 cutting edges for increased tool utility.
  • 11° relief angle for improved clearance.
  • Compatible with a broad range of materials including hardened steel, aluminum, brass, and titanium.
  • Suitable for applications in aerospace, medical, and mold making industries.
  • Design includes CPG geometry for medium application requirements.
Catalog No. 50-277-8668 Supplier Grainger Supplier No. 18D746
This diamond turning insert is designed for precision machining operations, including turning, boring, and profiling. Featuring a rhombic diamond shape, this uncoated insert is suitable for a wide range of workpiece materials, including steel, stainless steel, cast iron, nonferrous materials, and super alloys.
  • 4 cutting edges for increased tool utility.
  • 11° relief angle for improved clearance.
  • Compatible with a broad range of materials including hardened steel, aluminum, brass, and titanium.
  • Suitable for applications in aerospace, medical, and mold making industries.
  • Design includes CPG geometry for medium application requirements.