Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Precision Diamond Wafer Blade 6", Fine grit with High Concentration, 0.020" thick
SDP

Supplier:  Electron Microscopy Sciences 50265FH3

Encompass_Preferred

These bonded blades are constructed of an inner metal core and an outer rim, this rim is bonded with diamond particles. These blades come in high and low concentrations of diamond particles to handle various sectioning requirements. A high concentration of fine diamond particles is for general laboratory use with low or high saw's speed.
Particles: Fine
Concentration: High
Thickness: 0.020"

Catalog No. 50-997-194


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.