Learn More
Electron Microscopy Sciences Diamond Wafer Blade 4", 0.14" W, 0.20"D
SDP
Catalog No. 50997183
Encompass_Preferred
The blades are constructed of an inner metal core and an outer rim with diamond particles. This diamond wafering blade has a diameter of 4" and a thickness of 0.14" W, 0.20"D.
Catalog No. 50-997-183 Supplier Electron Microscopy Sciences Supplier No. 5026504
The blades are constructed of an inner metal core and an outer rim with diamond particles. This diamond wafering blade has a diameter of 4" and a thickness of 0.14" W, 0.20"D.