Learn More
Electron Microscopy Sciences Diamond Wafer Blade 4", 0.20" W, 0.20"D
SDP
Supplier:  Electron Microscopy Sciences 5026604
Encompass_Preferred
The blades are constructed of an inner metal core and an outer rim with diamond particles. This diamond wafering blade has a diameter of 4" and a thickness of .20"W .20"D.
Catalog No. 50-997-201