Learn More
Electron Microscopy Sciences Diamond Wafer Blade 6", 0.14" W, 0.20"D
SDP
Catalog No. 50997185
Encompass_Preferred
The blades are constructed of an inner metal core and an outer rim with diamond particles. This diamond wafering blade has a diameter of 6" and a thickness of 0.25" W, 0.276"D.
Catalog No. 50-997-185 Supplier Electron Microscopy Sciences Supplier No. 5026506
The blades are constructed of an inner metal core and an outer rim with diamond particles. This diamond wafering blade has a diameter of 6" and a thickness of 0.25" W, 0.276"D.