Learn More
Electron Microscopy Sciences Diamond Wafer Blade6", 0.20" W, 0.276"D
SDP
Catalog No. 50997211
Encompass_Preferred
The blades are constructed of an inner metal core and an outer rim with diamond particles. This diamond wafering blade has a diameter of 4" and a thickness of .32"W .276"D.
Catalog No. 50-997-211 Supplier Electron Microscopy Sciences Supplier No. 5026706
The blades are constructed of an inner metal core and an outer rim with diamond particles. This diamond wafering blade has a diameter of 4" and a thickness of .32"W .276"D.