Learn More
Electron Microscopy Sciences Diamond Wafer Blade 7", 0.25" W, 0.276"D
SDP
Catalog No. 50997186
Encompass_Preferred
The blades are constructed of an inner metal core and an outer rim with diamond particles. This diamond wafering blade has a diameter of 7" and a thickness of 0.14" W, 0.20"D.
Catalog No. 50-997-186 Supplier Electron Microscopy Sciences Supplier No. 5026507
The blades are constructed of an inner metal core and an outer rim with diamond particles. This diamond wafering blade has a diameter of 7" and a thickness of 0.14" W, 0.20"D.