Learn More
Heathrow Scientific Diamond Weigh Boats
GREENER_CHOICE
Catalog No. 01677178
Change view
Click to view available options
Capacity (Metric):
5 mL
25 mL
30 mL
100 mL
Color:
Black
White
Type:
Anti-static Weigh Boat
Standard Weigh Boat
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Color Type
01-677-178 5 mL White Standard Weigh Boat
01-677-174 5 mL White Anti-static Weigh Boat
01-677-182 5 mL Black Anti-static Weigh Boat
01-677-179 5 mL Black Standard Weigh Boat
01-677-169 25 mL White Standard Weigh Boat
01-677-184 25 mL White Anti-static Weigh Boat
01-677-181 25 mL Black Anti-static Weigh Boat
01-677-170 25 mL Black Standard Weigh Boat
01-677-185 30 mL White Anti-static Weigh Boat
01-677-173 30 mL Black Anti-static Weigh Boat
01-677-171 30 mL Black Standard Weigh Boat
01-677-175 100 mL White Standard Weigh Boat
01-677-180 100 mL White Anti-static Weigh Boat
01-677-183 100 mL Black Anti-static Weigh Boat
01-677-172 100 mL Black Standard Weigh Boat
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 missing translation for 'options'
Catalog No. 01-677-178 missing translation for 'mfr' Heathrow Scientific missing translation for 'supplierNo' HS1424AA
Add to Cart
Add to Cart
Ensures accurate sample measurements of liquids, granules, and powders
  • Smooth, flat surfaces with easy-pour spouts for precise sample handling
  • Rounded corners simplify complete sample transfer
  • Made from durable, single-use polystyrene for convenience and safety
  • FDA/food-grade compliant
  • Boats are shallow with a wide, flat bottom to resist tipping
  • Rounded corners simplify transfer
  • Available in the colors white and black for best color contrast
  • Can be used with microwave moisture analyzers
  • Disposable
  • Stackable
  • Anti-static versions available
  • Biologically inert and resistant to dilute acids, alcohols, and bases
Specifications
Bottom Shape Flat
Capacity (Metric) 5 mL
Color White
Material Polystyrene
Sterility Non-sterile
Type Standard Weigh Boat