Learn More
Analytical Sales & Services Diamond Weighing Boats - 100 mL
Catalog No. 5000807213
Encompass
Diamond Weighing Boats - 100mL • Diamond shape that easily flexes for accurate pouring • Wide, flat bottom to resist tipping L x W x H: 119 x 90 x 19 mm Color: White
Catalog No. 50-008-07213 Supplier Analytical Sales & Services Supplier No. DWB100
Diamond Weighing Boats - 100mL • Diamond shape that easily flexes for accurate pouring • Wide, flat bottom to resist tipping L x W x H: 119 x 90 x 19 mm Color: White