Learn More
Analytical Sales & Services Diamond Weighing Boats - 25 mL
Catalog No. 5000805297
Encompass
Diamond Weighing Boats - 25mL • Diamond shape that easily flexes for accurate pouring • Wide, flat bottom to resist tipping L x W x H: 71 x 46 x 14 mm Color: White
Catalog No. 50-008-05297 Supplier Analytical Sales & Services Supplier No. DWB25
Diamond Weighing Boats - 25mL • Diamond shape that easily flexes for accurate pouring • Wide, flat bottom to resist tipping L x W x H: 71 x 46 x 14 mm Color: White