Learn More
Analytical Sales & Services Diamond Weighing Boats - 30 mL
Catalog No. 5000805279
Encompass
Diamond Weighing Boats - 30mL • Diamond shape that easily flexes for accurate pouring • Wide, flat bottom to resist tipping L x W x H: 78 x 56 x 14 mm Color: White
Catalog No. 50-008-05279 Supplier Analytical Sales & Services Supplier No. DWB30
Diamond Weighing Boats - 30mL • Diamond shape that easily flexes for accurate pouring • Wide, flat bottom to resist tipping L x W x H: 78 x 56 x 14 mm Color: White