Learn More
Analytical Sales & Services Diamond Weighing Boats - 5 mL
Catalog No. 5000805324
Encompass
Diamond Weighing Boats - 5mL • Diamond shape that easily flexes for accurate pouring • Wide, flat bottom to resist tipping L x W x H: 55 x 35 x 6 mm Color: White
Catalog No. 50-008-05324 Supplier Analytical Sales & Services Supplier No. DWB5
Diamond Weighing Boats - 5mL • Diamond shape that easily flexes for accurate pouring • Wide, flat bottom to resist tipping L x W x H: 55 x 35 x 6 mm Color: White