Learn More
Checkers Diamondback™ Single Tunnel System
Catalog No. 19088418 Shop All Justrite Safety Group Products
Change view
Click to view available options
Color:
Yellow
Catalog No. Color
19-088-418 Yellow
1 options
Catalog No. 19-088-418 Supplier Checkers Supplier No. UHB5060
Only null left
Add to Cart
Add to Cart
Diamondback™ single tunnel systems provide protection for valuable cable and hose lines while ensuring a safe method of passage for vehicles. High-density, all-weather urethane construction protects from damage by physical pressure and reduces cable and hose maintenance and replacement costs.
  • Polyurethane Construction is Abrasion, Impact and Weather-Resistant with High Load-Bearing Capacity
  • Diamond-Plate Tread Surface Increases Traction
  • Yellow Caution Color for High Visibility
  • Modular Interlocking Design is Lightweight, Easy to Transport, Setup, Disassemble, and Store
  • Built-In Hand Holds Allow for Handling with Ease and Double as Attachment Points for Mounting Brackets
  • WARNING: Vehicles Crossing Bridge System Must Have Clearance of Greater Than Bridge Height
  • WARNING: Hose Bridges Are for Rubber Tired Vehicles Only (Not for Equipment with Metal Tracks)
Specifications
Color Yellow
Material Polyurethane
Product Type Cable Protector