Learn More
DSI LLC DIAMONDBOND - C18 REVERSED-PH
Supplier:  DSI LLC DB011021
Encompass_Preferred
DiamondBond-C18 eliminates the shortcomings of conventional ODS-silica columns by combining patented surface technology with ZirChrom's state-of-the-art particles. Carbon-clad zirconia, stable at all levels of pH.
Catalog No. 50-123-044
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.