Learn More
Diamonite™ Mortar and Pestle Sets
Synthetic sapphire mortars offer excellent hardness and chemical resistance
Manufacturer:  Diamonite Products ManufacturingSupplier Diversity Partner MP65
 View more versions of this product
Catalog No. 12-952
  Check Availability
Add to cart
Alert:
Not recommended for cryogenic use.
Specifications Description & Specifications
Specifications
2.2 in.
6.5cm
Mortar and Pestle
For grinding or crushing
4cm
3.4 in.
8.5cm
Diamonite
1.6 in.