Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Diamthazole hydrochloride | 17140-69-1 | 99.9% | 329.89 | C15H24ClN3OS | 10 MG
SDP

Catalog No. 5000286700
Encompass_Preferred

Diamthazole hydrochloride (Dimazole hydrochloride) is an antifungal compound used in infection research for in vitro and in vivo studies. It is supplied as a solid powder with documented solubility protocols and storage recommendations for research use.

  • Molecular formula: C15H24ClN3OS.
  • CAS number: 17140-69-1.
  • Molecular weight: 329.89.
  • Purity: 99.86% (reported).
  • Appearance: white to light yellow solid.
  • Solubility: DMSO 100 mg/mL; in vivo formulations 2.5 mg/mL in several vehicle systems (requires ultrasonic or warming).
  • Storage: solid at 4°C sealed; in solvent -80°C (6 months) or -20°C (1 month).
  • Available pack sizes: 5 mg, 10 mg, 25 mg, 50 mg, 100 mg (quote), 200 mg (quote).

Catalog No. 50-002-86700 Supplier Medchemexpress LLC Supplier No. HY148160A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Diamthazole hydrochloride (Dimazole hydrochloride) is an antifungal compound used in infection research for in vitro and in vivo studies. It is supplied as a solid powder with documented solubility protocols and storage recommendations for research use.

  • Molecular formula: C15H24ClN3OS.
  • CAS number: 17140-69-1.
  • Molecular weight: 329.89.
  • Purity: 99.86% (reported).
  • Appearance: white to light yellow solid.
  • Solubility: DMSO 100 mg/mL; in vivo formulations 2.5 mg/mL in several vehicle systems (requires ultrasonic or warming).
  • Storage: solid at 4°C sealed; in solvent -80°C (6 months) or -20°C (1 month).
  • Available pack sizes: 5 mg, 10 mg, 25 mg, 50 mg, 100 mg (quote), 200 mg (quote).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.