Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diamyl Phthalate, SPEX CertiPrep™
SDP

Catalog No. S4157 Shop All Spex CertiPrep Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S4157 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S4157 Supplier SPEX CertiPrep Supplier No. S4157

missing translation for 'validMainframeMsg'

Comprend

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
Diamyl phthalate131-18-01000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Identifiants chimiques

CAS 131-18-0
Molecular Formula C18H26O4
Molecular Weight (g/mol) 306.402
InChI Key IPKKHRVROFYTEK-UHFFFAOYSA-N
PubChem CID 8561
ChEBI CHEBI:34680
IUPAC Name dipentyl benzene-1,2-dicarboxylate
SMILES CCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCC

Spécifications

Packaging 2mL amber prescored glass ampule
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Solvent or Matrix Methanol-P and T
Quantity 1 mL
Concentration or Composition (by Analyte or Components) 1000μg/mL
Chemical Name or Material Diamyl phthalate
DOTInformation Nonhazardous
Recommended Storage Ambient

Product intended for laboratory use only

missing translation for 'productTitle'
missing translation for 'pleaseSelectIssue'

missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.