Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diamyl Sulfite 97.0+%, TCI America™
SDP

Catalog No. D297525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D297525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D297525G Supplier TCI America Supplier No. D297525G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2051-05-0
Molecular Formula C10H22O3S
Molecular Weight (g/mol) 222.34
MDL Number MFCD06252141
InChI Key QHKKGWHSXSIVAJ-UHFFFAOYSA-N
Synonym Dipentyl Sulfite
PubChem CID 137299
IUPAC Name dipentyl sulfite
SMILES CCCCCOS(=O)OCCCCC

Specifications

Color Colorless
Quantity 25 g
Formula Weight 222.34
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Diamyl Sulfite
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.