Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diamylamine (mixture) 97.0+%, TCI America™
SDP

Catalog No. D0286500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0286500ML 500 mL
D028625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0286500ML Supplier TCI America Supplier No. D0286500ML

Chemical Identifiers

CAS 2050-92-2
Molecular Formula C10H23N
Molecular Weight (g/mol) 157.301
MDL Number MFCD00009499
InChI Key JACMPVXHEARCBO-UHFFFAOYSA-N
Synonym dipentylamine, diamylamine, di-n-amylamine, 1-pentanamine, n-pentyl, di-n-pentylamine, diamyl amine, pentylamine, pentyl, amine, dipentyl, unii-d67i8pn8uu, ccris 6225
PubChem CID 16316
IUPAC Name N-pentylpentan-1-amine
SMILES CCCCCNCCCCC

Specifications

Color Yellow
Quantity 500 mL
UN Number 1992
Formula Weight 157.30
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Diamylamine (mixture)
RTECSNumber RZ9100000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.