Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diamylamine 98.0+%, TCI America™
SDP

Catalog No. D012925ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D012925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D012925ML Supplier TCI America Supplier No. D012925ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2050-92-2
Molecular Formula C10H23N
Molecular Weight (g/mol) 157.301
MDL Number MFCD00009499
InChI Key JACMPVXHEARCBO-UHFFFAOYSA-N
Synonym dipentylamine, diamylamine, di-n-amylamine, 1-pentanamine, n-pentyl, di-n-pentylamine, diamyl amine, pentylamine, pentyl, amine, dipentyl, unii-d67i8pn8uu, ccris 6225
PubChem CID 16316
IUPAC Name N-pentylpentan-1-amine
SMILES CCCCCNCCCCC

Specifications

Melting Point -8°C
Color Yellow
Boiling Point 202°C
Quantity 25 mL
UN Number 2841
Formula Weight 157.30
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diamylamine
EINECSNumber (2)-3149
RTECSNumber RZ9100000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.