Learn More
StatLab Diapath Dante Cold Plate Module
SDP
Catalog No. 21402206
Change view
Click to view available options
Product Type:
Cold Plate Module
Catalog No. Product Type
21-402-206 Cold Plate Module
1 options
Catalog No. 21-402-206 Supplier StatLab Supplier No. SDSDF9100
Add to Cart
Add to Cart
Efficient cold plate module designed for rapid cooling and solidification of embedded samples in histology laboratories.
  • Cold Plate Module
  • For Use with the StatLab Diapath Dante Embedding Center
  • 1 Ea.
Specifications
Product Type Cold Plate Module