Learn More
StatLab Diapath Dante Embedding Center Kit
SDP
Catalog No. 21402196
Change view
Click to view available options
Slide Capacity:
126 Casettes
Catalog No. Slide Capacity
21-402-196 126 Casettes
1 options
Catalog No. 21-402-196 Supplier StatLab Supplier No. DANTEECKIT
Add to Cart
Add to Cart
Diapath Dante Embedding Center Kit: Maximize comfort and efficiency - For labs that value performance and comfort, experience a smart embedding solution built to improve lab workflow and your well-being.
  • StatLab Diapath Dante Embedding Center Kit Includes Embedding Center Module Cold Plate Magnifying Glass with LED Lamp Paraffin Dispensing Foot Pedal
  • Modular System Consists of an embedding module and a cold plate fully flexible for multiple solutions
  • Large 6 L Paraffin Tank Paraffin can be melted with a temperature range between +50°C to +75°C
  • Wide Heated Surface Features precision grooves designed for efficient paraffin drainage
  • LED Light The work area is illuminated by an adjustable LED light integrated in the dispensing head
  • Intuitive Software Alerts operators when the paraffin tank needs to be filled minimizing work interruptions and downtime
  • Wide Cold Plate Fits up to 126 cassettes almost double the capacity of other models available
  • Ultrasonic Sensor System can dispense paraffin upon detecting the presence of a mold
  • Condensation Channel Collects drips from defrosting in a channel around the edge of the instrument
  • Removable Forceps Holder for easy cleaning
Specifications
Product Type Embedding Center
Slide Capacity 126 Casettes