Learn More
StatLab Diapath Galileo Semi-Automated Microtome With Standard Clamp Head Kit
SDP
Catalog No. 21402200
Change view
Click to view available options
Profile:
Low
Catalog No. Profile
21-402-200 Low
1 options
Catalog No. 21-402-200 Supplier StatLab Supplier No. GALISEMIS2EULPSTDKIT
Add to Cart
Add to Cart
Diapath Galileo Semi-Automated Microtome With Standard Clamp Head Kit: Flexible, reliable, precise - Advanced customization and innovative features to meet the demands of busy labs.
  • StatLab Diapath Galileo Semi-Automated Microtome with Standard Clamp Head Kit: Includes Microtome, Standard Adjustable Head, Universal Low profile Blade Holder, Quick Release Clamp
  • Ergonomic - with intuitive controls and ergonomic design, it ensures precision, reduces strain, and enhances productivity
  • Reliable - Engineered for long-lasting performance, this microtome is reliable over the years thanks to the robust design
  • Customizable - Adapts to your needs with interchangeable blade holders, precision clamps, and memory functions, for optimal performance
  • Efficient - Ensures efficiency in demanding lab environments, minimizing downtime even in tough conditions
  • Optional PRO System for higher throughput and traceability
  • Manufactured in Italy in an ISO-13485 certified facility
  • The microtomes can be used with histology equipments.
Specifications
Product Type Microtome
Profile Low