Learn More
Worthington Biochemical Corporation DIAPHORASE 1 KU
SDP
Catalog No. NC9210060
Encompass
Diaphorase, Code: DILW, Store at 2 deg. to 8 deg.C, Minimum activity: 30U/mg dry weight, Supplied as a dialyzed, lyophilized powder, Unit Definition: 1 Unit equals a decrease in absorbance at 600nm of 1.0 per minute at 25 deg.C, pH 7.5, 1kU
Catalog No. NC9210060 Supplier Worthington Biochemical Corporation Supplier No. LS004327
Diaphorase, Code: DILW, Store at 2 deg. to 8 deg.C, Minimum activity: 30U/mg dry weight, Supplied as a dialyzed, lyophilized powder, Unit Definition: 1 Unit equals a decrease in absorbance at 600nm of 1.0 per minute at 25 deg.C, pH 7.5, 1kU