Learn More
Enzo Life Sciences Diaphorase (500U). CAS: 9001-68-7
SDP
Catalog No. 502013944
Encompass_Preferred
Alternative name: NADPH dehydrogenase. Purity: ≥95%. Formulation: Liquid. In 50mM TRIS, pH 8.0, containing 150mM NaCl. Source: Produced in E. coli. Long Term Storage: -80°C.
Catalog No. 50-201-3944 Supplier Enzo Life Sciences Supplier No. ENZPRT1550500
Alternative name: NADPH dehydrogenase. Purity: ≥95%. Formulation: Liquid. In 50mM TRIS, pH 8.0, containing 150mM NaCl. Source: Produced in E. coli. Long Term Storage: -80°C.