Learn More
Grainger Diaphragm Assembly Toilets
Catalog No. 502766427
Encompass_Preferred
The Sloan Diaphragm Assembly is designed to fit Sloan flush valves including the Royal 111 and 186 models. It is made of neoprene rubber and measures 3-1/2" in diameter. This diaphragm assembly acts as a seal between the upper and lower portion of the flush valve, closing off the water supply. It features a center hole to allow the flush valve plunger to pass through. The diaphragm should be replaced when it becomes cracked, warped, or fails to make a tight seal. This is a genuine OEM replacement part from Sloan that will restore proper function to compatible flush valves.
Catalog No. 50-276-6427 Supplier Grainger Supplier No. 16U047
The Sloan Diaphragm Assembly is designed to fit Sloan flush valves including the Royal 111 and 186 models. It is made of neoprene rubber and measures 3-1/2" in diameter. This diaphragm assembly acts as a seal between the upper and lower portion of the flush valve, closing off the water supply. It features a center hole to allow the flush valve plunger to pass through. The diaphragm should be replaced when it becomes cracked, warped, or fails to make a tight seal. This is a genuine OEM replacement part from Sloan that will restore proper function to compatible flush valves.