Learn More
Grainger Diaphragm Assembly Toilets
Catalog No. 502749714
Encompass_Preferred
The Kissler Diaphragm Assembly is a replacement part for Coyne & Delany Series 1400 and 1700 liquid pumps. It has a 316 stainless steel body with EPDM diaphragms and is designed for use with compatible pumps from 1/4" to 2" discharge sizes. The assembly measures 5.75" diameter x 3.25" thick and weighs approximately 3 lbs. It has a maximum pressure rating of 250 PSI and temperature range of -20 to 180 degrees Fahrenheit. The Kissler diaphragm assembly is an OEM replacement part made to match the original Coyne & Delany pump specifications for fit and performance.
Catalog No. 50-274-9714 Supplier Grainger Supplier No. 14U340
The Kissler Diaphragm Assembly is a replacement part for Coyne & Delany Series 1400 and 1700 liquid pumps. It has a 316 stainless steel body with EPDM diaphragms and is designed for use with compatible pumps from 1/4" to 2" discharge sizes. The assembly measures 5.75" diameter x 3.25" thick and weighs approximately 3 lbs. It has a maximum pressure rating of 250 PSI and temperature range of -20 to 180 degrees Fahrenheit. The Kissler diaphragm assembly is an OEM replacement part made to match the original Coyne & Delany pump specifications for fit and performance.