Learn More
Chemglass Life Sciences Diaphragm Liquid Metering Pump, 1-100mL/Min, Compressions for soft tubing 4mm ID x 6mm OD
SDP
Catalog No. 501311012
Encompass_Preferred
Diaphragm Liquid Metering Pump, 1-100mL/Min, Compressions for soft tubing 4mm ID x 6mm OD. The easy-to-read display combined w/ a touch control knob makes programming & access of all functions very intuitive. Diaphragm pumps are ideally suited for transferring the corrosive liquids often found in laboratories.The SIMDOS pump wetted materials are PVDF/PTFE/FFKM. The pump head is situated outside the splash-protected IP-65 housing, a significant safety factor.Following calibration, repeatability is +/-1%. There are two different model options for transfer rates with a flow rate of .02-30mL and 1-100mL/min. They allow doses of volumes of 0 to 1000mL. The pump is safe to run dry & is self-priming up to 9ft of water. It is effective against pressures of up to 88 psig & transfers viscous media up to 150 centistokes. Uses compression fittings for tubing.
Catalog No. 50-131-1012 Supplier Chemglass Life Sciences Supplier No. CG1170P100
Diaphragm Liquid Metering Pump, 1-100mL/Min, Compressions for soft tubing 4mm ID x 6mm OD. The easy-to-read display combined w/ a touch control knob makes programming & access of all functions very intuitive. Diaphragm pumps are ideally suited for transferring the corrosive liquids often found in laboratories.The SIMDOS pump wetted materials are PVDF/PTFE/FFKM. The pump head is situated outside the splash-protected IP-65 housing, a significant safety factor.Following calibration, repeatability is +/-1%. There are two different model options for transfer rates with a flow rate of .02-30mL and 1-100mL/min. They allow doses of volumes of 0 to 1000mL. The pump is safe to run dry & is self-priming up to 9ft of water. It is effective against pressures of up to 88 psig & transfers viscous media up to 150 centistokes. Uses compression fittings for tubing.