Learn More
Grainger Diaphragm Pump Flange Kit PP.1-1/2 In.
Catalog No. 502765511
Encompass_Preferred
The Grainger Approved Diaphragm Pump Flange Kit is designed to work with Wilden diaphragm pumps. It includes 4 flanges, 8 gaskets, and 16 bolts and nuts to allow you to install the pump. The flanges are made of polypropylene for chemical resistance. The gaskets are Buna-N for compatibility with water-based fluids. The bolts are zinc plated steel and the nuts are zinc plated steel nylon insert locknuts. This kit will let you install a 1/2", 3/4", or 1" Wilden diaphragm pump. It is ideal for water transfer applications.
Catalog No. 50-276-5511 Supplier Grainger Supplier No. 16X839
The Grainger Approved Diaphragm Pump Flange Kit is designed to work with Wilden diaphragm pumps. It includes 4 flanges, 8 gaskets, and 16 bolts and nuts to allow you to install the pump. The flanges are made of polypropylene for chemical resistance. The gaskets are Buna-N for compatibility with water-based fluids. The bolts are zinc plated steel and the nuts are zinc plated steel nylon insert locknuts. This kit will let you install a 1/2", 3/4", or 1" Wilden diaphragm pump. It is ideal for water transfer applications.