Learn More
Molecular Dimensions DIAPLATE 96 SAMPLE DIALYSIS(3)
SDP
Catalog No. 501038648
Encompass
Diaplate 96 Sample 3.2uL Dialysis Plate 10kDa MWCO Pack of 3
Catalog No. 50-103-8648 Supplier Molecular Dimensions Supplier No. MD670
Diaplate 96 Sample 3.2uL Dialysis Plate 10kDa MWCO Pack of 3