Learn More
Medchemexpress LLC Diastase, Aspergillus oryzae | 9000-92-4 | 10 G
SDP
Catalog No. 5000412404
Encompass_Preferred
Diastase is an amylase from Aspergillus oryzae that hydrolyzes starch into sugars.
  • Enzyme activity: ≥100000 U/g
  • Unit definition: One unit of enzyme activity is defined as the amount of enzyme that can liquefy 1 g of soluble at 40°C and pH 5.0 in 1 hour.
  • For research use only.
  • We do not sell to patients.
Catalog No. 50-004-12404 Supplier Medchemexpress LLC Supplier No. HYB2192A10G
Diastase is an amylase from Aspergillus oryzae that hydrolyzes starch into sugars.
  • Enzyme activity: ≥100000 U/g
  • Unit definition: One unit of enzyme activity is defined as the amount of enzyme that can liquefy 1 g of soluble at 40°C and pH 5.0 in 1 hour.
  • For research use only.
  • We do not sell to patients.