Learn More
Medchemexpress LLC Diastase, Aspergillus oryzae | 9000-92-4 | 5 G
SDP
Catalog No. 5000410353
Encompass_Preferred
Diastase from Aspergillus oryzae is an amylase that hydrolyzes starch into sugars. This product is intended for research use only and has not been fully validated for medical applications.
  • Enzyme activity of ≥100000 U/g.
  • Appears as a solid, light yellow to yellow powder.
  • Hydrolyzes starch into sugars.
  • Suitable for laboratory chemical applications.
Catalog No. 50-004-10353 Supplier Medchemexpress LLC Supplier No. HYB2192A5G
Diastase from Aspergillus oryzae is an amylase that hydrolyzes starch into sugars. This product is intended for research use only and has not been fully validated for medical applications.
  • Enzyme activity of ≥100000 U/g.
  • Appears as a solid, light yellow to yellow powder.
  • Hydrolyzes starch into sugars.
  • Suitable for laboratory chemical applications.