Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diastase from Malt , MP Biomedicals, LLC

Catalog No. MP210153991 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Plastic
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MP210153991 1 kg Plastic
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP210153991 Supplier MP Biomedicals, Inc Supplier No. 0210153991
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Diastase from Malt 1X N.F. Stable, 60 mesh powder.

1g will digest 50g starch in 30 minutes
TRUSTED_SUSTAINABILITY

Specifications

Color White
Packaging Plastic
Quantity 1 kg
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.