Learn More
AdipoGen Diastovaricin I
SDP
Supplier:  AdipoGen BVT0381M005
Encompass_Preferred
Chemical. CAS 102281-52-7. Formula C39H45NO10. MW 687.8. Isolated from Streptomyces sp. Gö 40/10. Antibiotic. Antibacterial. Inducer of Friend mouse erythroleukemia cells. Antitumor compound.
Catalog No. 50-256-1962