Learn More
Sigma Aldrich Fine Chemicals Biosciences Celite(R) 503 filter aid, treated with sodium carbonate, calcined | 68855-54-9 | MFCD00132803 | 1KG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 221511KG
Encompass
Celite(R) 503 filter aid, treated with sodium carbonate, calcined | 68855-54-9 | MFCD00132803 | 1KG
Catalog No. 50-187-9629