Learn More
Electron Microscopy Sciences DiATOME Histo Jumbo Diamond Knife 8.0 mm, 45° Knife angle
SDP
Catalog No. 501900064
Encompass
The large Jumbo boat as well as the adhesive (Pattex compact by Henkel) applied to the side of the sample block increase the distinct advantages. They allow easy production of section ribbons (0.5-2 micrometers), no section loss, no folding, and the same orientation of all sections. As well as easy collection of section ribbons, multiple ribbons on one glass slide, and perfect for immuno-histo-chemistry.
Size: 8.0 mm
Knife angle: 45°
Catalog No. 50-190-0064 Supplier Electron Microscopy Sciences Supplier No. 80HISJ
Add to Cart
Add to Cart
Request a Quote
The large Jumbo boat as well as the adhesive (Pattex compact by Henkel) applied to the side of the sample block increase the distinct advantages. They allow easy production of section ribbons (0.5-2 micrometers), no section loss, no folding, and the same orientation of all sections. As well as easy collection of section ribbons, multiple ribbons on one glass slide, and perfect for immuno-histo-chemistry.
Size: 8.0 mm
Knife angle: 45°