Learn More
Electron Microscopy Sciences DiATOME manip
SDP
Catalog No. 501926572
Encompass
For easily handling and mounting of section ribbons on TEM support film grids. To use the original plate on the cryochamber is removed, the manipulator is fixed in 5 minutes.
Applications include frozen hydrated biological samples (CEMOVIS) and room temperature sectioning of water sensitive samples such as biological samples for chemical analysis or polymers.
The left manipulator moves a conductive fibre on which the sections are attached by electrostatic force; the growing ribbon is guided. The right manipulator guides the grid very precisely below the section ribbon, the sections are attached to the carbon film with electrostatic force.
Catalog No. 50-192-6572 Supplier Electron Microscopy Sciences Supplier No. DZ10001
Add to Cart
Add to Cart
Request a Quote
For easily handling and mounting of section ribbons on TEM support film grids. To use the original plate on the cryochamber is removed, the manipulator is fixed in 5 minutes.
Applications include frozen hydrated biological samples (CEMOVIS) and room temperature sectioning of water sensitive samples such as biological samples for chemical analysis or polymers.
The left manipulator moves a conductive fibre on which the sections are attached by electrostatic force; the growing ribbon is guided. The right manipulator guides the grid very precisely below the section ribbon, the sections are attached to the carbon film with electrostatic force.