Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Diatrizoic Acid, 100 g

Catalog No. ICN19150180 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19150180 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19150180 Supplier MP Biomedicals™ Supplier No. 0219150180

missing translation for 'validMainframeMsg'

Diatrizoic acid is commonly used as a contrast agent in radiography.

  • Very slightly soluble in water and in alcohol
  • Soluble in dimethylformamide and in alkali hydroxide solutions
TRUSTED_SUSTAINABILITY

Specifications

CAS 50978-11-5
Quantity 100 g

Unless specified otherwise, MP Biomedical™'s products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.